siRNA Details
| Gene Symbol: Tmem119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146162.2 | NP_666274.1 | 2 | 1700 | ||
| GenBank mRNA | AK078473.1 | - | 2 | 1709 | ||
| AK170712.1 | - | 2 | 1710 | |||
| BC025600.1 | - | 2 | 1705 | 5250899 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem119| Gene Symbol: Tmem119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146162.2 | NP_666274.1 | 2 | 1700 | ||
| GenBank mRNA | AK078473.1 | - | 2 | 1709 | ||
| AK170712.1 | - | 2 | 1710 | |||
| BC025600.1 | - | 2 | 1705 | 5250899 | ||