siRNA Details
| Gene Symbol: Tspan33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301407.1 | NP_001288336.1 | 8 | 1294 | ||
| NM_146173.3 | NP_666285.1 | 8 | 1297 | |||
| GenBank mRNA | AF276890.1 | - | 8 | 1296 | ||
| AK004954.1 | - | 8 | 1270 | |||
| AK172176.1 | - | 8 | 1275 | |||
| BC024685.1 | - | 8 | 1255 | 3981492 | ||

Tspan33
Genomic Map