siRNA Details
| Gene Symbol: Zfp282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146175.3 | NP_666287.3 | 8 | 2485 | ||
| GenBank mRNA | AK030516.1 | - | 8 | 2393 | ||
| AK030919.1 | - | 1 | 937 | |||
| AK088531.1 | - | 8 | 2501 | |||
| AK148038.1 | - | 8 | 2472 | |||
| BC025581.1 | - | 9 | 2007 | 5325029 | ||

Zfp282
Genomic Map