siRNA Details
| Gene Symbol: Zfp790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145880.1 | NP_001139352.1 | 5 | 2865 | ||
| NM_146185.2 | NP_666297.2 | 5 | 2955 | |||
| GenBank mRNA | AK018258.1 | - | 1 | 208 | ||
| AK030519.1 | - | 5 | 2956 | |||
| AK031224.1 | - | 1 | 709 | |||
| BC007476.1 | - | 5 | 2908 | 3497180 | ||
| BC046774.1 | - | 5 | 2823 | 5708351 | ||

Zfp790
Genomic Map