siRNA Details
| Gene Symbol: Eif3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146200.1 | NP_666312.1 | 21 | 2728 | ||
| GenBank mRNA | AK088076.1 | - | 21 | 2772 | ||
| AK152693.1 | - | 21 | 2746 | |||
| AK160078.1 | - | 21 | 2748 | |||
| AK168160.1 | - | 21 | 2732 | |||
| AK168174.1 | - | 21 | 2732 | |||
| BC024855.1 | - | 21 | 2728 | 5359060 | ||
| BC025032.1 | - | 21 | 2749 | 4947435 | ||
| BC080763.1 | - | 8 | 1143 | |||

Eif3c
Genomic Map