siRNA Details
| Gene Symbol: CSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000759.3 | NP_000750.1 | 5 | 1578 | ||
| NM_001178147.1 | NP_001171618.1 | 4 | 1461 | |||
| NM_172219.2 | NP_757373.1 | 5 | 1569 | |||
| NM_172220.2 | NP_757374.2 | 4 | 1470 | |||
| NR_033662.1 | 4 | 1745 | ||||
| GenBank mRNA | BC033245.1 | - | 4 | 1639 | 5434958 | |
| X03438.1 | - | 5 | 1495 | |||

CSF3
Genomic Map