siRNA Details
| Gene Symbol: Fam199x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146261.2 | NP_666373.1 | 2 | 536 | ||
| GenBank mRNA | AK039695.1 | - | 2 | 464 | ||
| AK154624.1 | - | 2 | 539 | |||
| BC031748.1 | - | 2 | 501 | 3672969 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam199x| Gene Symbol: Fam199x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146261.2 | NP_666373.1 | 2 | 536 | ||
| GenBank mRNA | AK039695.1 | - | 2 | 464 | ||
| AK154624.1 | - | 2 | 539 | |||
| BC031748.1 | - | 2 | 501 | 3672969 | ||