siRNA Details
| Gene Symbol: Gm13769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270423.1 | NP_001257352.1 | 2 | 634 | ||
| GenBank mRNA | CB174322.1 | - | 1 | 468 | ||
| Gene Symbol: Olfr1506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146265.2 | NP_666377.2 | 2 | 634 | ||

Gm13769
Genomic Map