siRNA Details
| Gene Symbol: Olfr930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146272.1 | NP_666384.1 | 1 | 341 | ||
| GenBank mRNA | BC119252.1 | - | 1 | 423 | 8734001 | |
| BC119254.1 | - | 1 | 425 | 8734003 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr930| Gene Symbol: Olfr930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146272.1 | NP_666384.1 | 1 | 341 | ||
| GenBank mRNA | BC119252.1 | - | 1 | 423 | 8734001 | |
| BC119254.1 | - | 1 | 425 | 8734003 | ||