siRNA Details
Gene Symbol: Olfr1311 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146274.1 | NP_666386.1 | 1 | 294 | ||
GenBank mRNA | BC127967.1 | - | 1 | 298 | 40135022 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1311 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146274.1 | NP_666386.1 | 1 | 294 | ||
GenBank mRNA | BC127967.1 | - | 1 | 298 | 40135022 |