siRNA Details
Gene Symbol: Olfr1412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146277.1 | NP_666389.1 | 1 | 359 | ||
GenBank mRNA | BC125241.1 | - | 1 | 359 | 40044004 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146277.1 | NP_666389.1 | 1 | 359 | ||
GenBank mRNA | BC125241.1 | - | 1 | 359 | 40044004 |