siRNA Details
| Gene Symbol: Olfr281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146280.1 | NP_666392.1 | 1 | 257 | ||
| GenBank mRNA | BC119146.1 | - | 1 | 284 | 8733895 | |
| BC120834.1 | - | 1 | 284 | 40129757 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr281| Gene Symbol: Olfr281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146280.1 | NP_666392.1 | 1 | 257 | ||
| GenBank mRNA | BC119146.1 | - | 1 | 284 | 8733895 | |
| BC120834.1 | - | 1 | 284 | 40129757 | ||