siRNA Details
| Gene Symbol: Olfr780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146284.1 | NP_666396.1 | 1 | 419 | ||
| GenBank mRNA | BC138405.1 | - | 1813 | 8861425 | ||
| BC138406.1 | - | 1813 | 8861426 | |||
| BC147436.1 | - | 1 | 475 | 9087435 | ||

Olfr780
Genomic Map