siRNA Details
| Gene Symbol: Olfr727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146319.2 | NP_666431.2 | 1 | 964 | ||
| GenBank mRNA | BC132575.1 | - | 1 | 964 | 40130852 | |
| BC145712.1 | - | 1 | 964 | 40131015 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr727| Gene Symbol: Olfr727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146319.2 | NP_666431.2 | 1 | 964 | ||
| GenBank mRNA | BC132575.1 | - | 1 | 964 | 40130852 | |
| BC145712.1 | - | 1 | 964 | 40131015 | ||