siRNA Details
| Gene Symbol: Olfr186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146321.1 | NP_666433.1 | 1 | 629 | ||
| GenBank mRNA | BC150713.1 | - | 1 | 651 | 9087624 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr186| Gene Symbol: Olfr186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146321.1 | NP_666433.1 | 1 | 629 | ||
| GenBank mRNA | BC150713.1 | - | 1 | 651 | 9087624 | |