siRNA Details
| Gene Symbol: Olfr958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146330.1 | NP_666442.1 | 1 | 880 | ||
| GenBank mRNA | BC148230.1 | - | 1 | 884 | 40142225 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr958| Gene Symbol: Olfr958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146330.1 | NP_666442.1 | 1 | 880 | ||
| GenBank mRNA | BC148230.1 | - | 1 | 884 | 40142225 | |