siRNA Details
| Gene Symbol: FOLR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000803.4 | NP_000794.3 | 4, 5 | 658 | ||
| NM_001113534.1 | NP_001107006.1 | 4, 5 | 652 | |||
| NM_001113535.1 | NP_001107007.1 | 4, 5 | 646 | |||
| NM_001113536.1 | NP_001107008.1 | 4, 5 | 640 | |||
| XM_005273856.3 | XP_005273913.1 | 4, 5 | 656 | |||
| GenBank mRNA | AK222539.1 | - | 4, 5 | 649 | ||
| AK222643.1 | - | 4, 5 | 640 | |||
| BC027894.1 | - | 4, 5 | 627 | |||
| BC058036.1 | - | 4, 5 | 642 | 6662871 | ||
| BC115366.1 | - | 3, 4 | 428 | |||
| CD014063.1 | - | 3, 4 | 509 | |||
| CD692073.1 | - | 4, 5 | 656 | |||

FOLR2
Genomic Map