siRNA Details
| Gene Symbol: Cdk17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146239.2 | NP_666351.2 | 4, 5 | 779 | ||
| GenBank mRNA | AK078325.1 | - | 4, 5 | 780 | ||
| BC031778.1 | - | 326 | 4501115 | |||
| BC049904.1 | - | 326 | 4501115 | |||
| BC064815.1 | - | 4, 5 | 764 | 6516981 | ||
| CD350146.1 | - | 3, 4 | 409 | |||

Cdk17
Genomic Map