siRNA Details
| Gene Symbol: Lrrc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146242.2 | NP_666354.1 | 1 | 924 | ||
| GenBank mRNA | AB091100.1 | - | 1 | 894 | ||
| AF527781.1 | - | 1 | 910 | |||
| AK084466.1 | - | 1 | 909 | |||
| AK085779.1 | - | 1 | 912 | |||
| AK142167.1 | - | 1 | 911 | |||
| BC086911.1 | - | 1 | 924 | 6752714 | ||

Lrrc10
Genomic Map