siRNA Details
| Gene Symbol: Olfr1402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146275.1 | NP_666387.1 | 1 | 663 | ||
| GenBank mRNA | BC108950.1 | - | 1 | 663 | 40044400 | |
| BC108951.1 | - | 1 | 663 | 40044401 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1402| Gene Symbol: Olfr1402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146275.1 | NP_666387.1 | 1 | 663 | ||
| GenBank mRNA | BC108950.1 | - | 1 | 663 | 40044400 | |
| BC108951.1 | - | 1 | 663 | 40044401 | ||