siRNA Details
| Gene Symbol: Olfr1366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146283.2 | NP_666395.2 | 1 | 189 | ||
| GenBank mRNA | BC138101.1 | - | 1 | 187 | 8861120 | |
| BC138102.1 | - | 1 | 189 | 8861121 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1366| Gene Symbol: Olfr1366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146283.2 | NP_666395.2 | 1 | 189 | ||
| GenBank mRNA | BC138101.1 | - | 1 | 187 | 8861120 | |
| BC138102.1 | - | 1 | 189 | 8861121 | ||