siRNA Details
| Gene Symbol: GGCX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000821.6 | NP_000812.2 | 15 | 2644 | ||
| NM_001142269.3 | NP_001135741.1 | 14 | 2473 | |||
| XM_005264259.4 | XP_005264316.1 | 15 | 2597 | |||
| XM_011532764.2 | XP_011531066.1 | 14 | 2439 | |||
| XM_011532765.2 | XP_011531067.1 | 13 | 2278 | |||
| XM_017003803.1 | XP_016859292.1 | 14 | 2460 | |||
| GenBank mRNA | AK130139.1 | - | 6 | 1235 | ||
| AK298982.1 | - | 14 | 2298 | |||
| BC013979.2 | - | 15 | 2566 | 4129593 | ||

GGCX
Genomic Map