siRNA Details
| Gene Symbol: Olfr827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146300.1 | NP_666412.1 | 1 | 457 | ||
| GenBank mRNA | BC117109.1 | - | 1 | 478 | 40126428 | |
| BC117111.1 | - | 1 | 478 | 40126430 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr827| Gene Symbol: Olfr827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146300.1 | NP_666412.1 | 1 | 457 | ||
| GenBank mRNA | BC117109.1 | - | 1 | 478 | 40126428 | |
| BC117111.1 | - | 1 | 478 | 40126430 | ||