siRNA Details
| Gene Symbol: Olfr642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146329.1 | NP_666441.1 | 1 | 167 | ||
| GenBank mRNA | BC126862.1 | - | 1 | 167 | 40042372 | |
| BC126863.1 | - | 1 | 167 | 40042373 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr642| Gene Symbol: Olfr642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146329.1 | NP_666441.1 | 1 | 167 | ||
| GenBank mRNA | BC126862.1 | - | 1 | 167 | 40042372 | |
| BC126863.1 | - | 1 | 167 | 40042373 | ||