siRNA Details
Gene Symbol: Olfr374 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146338.2 | NP_666450.2 | 2 | 417 | ||
GenBank mRNA | BC145662.1 | - | 1 | 361 | 40130965 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr374 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146338.2 | NP_666450.2 | 2 | 417 | ||
GenBank mRNA | BC145662.1 | - | 1 | 361 | 40130965 |