siRNA Details
| Gene Symbol: Tmie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146260.2 | NP_666372.1 | 5, 6 | 1535 | ||
| GenBank mRNA | AF481143.1 | - | 5, 6 | 1535 | ||
| AK030341.1 | - | 5, 6 | 1565 | |||
| AK031966.1 | - | 5, 6 | 1597 | |||
| BC138840.1 | - | 5, 6 | 1099 | 8861862 | ||
| BC141124.1 | - | 5, 6 | 1099 | 9055736 | ||

Tmie
Genomic Map