siRNA Details
| Gene Symbol: Olfr1506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146265.2 | NP_666377.2 | 2 | 1036 | ||
| GenBank mRNA | BC111438.1 | - | 3 | 515 | ||
| Gene Symbol: Gm13769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270423.1 | NP_001257352.1 | 2 | 1036 | ||

Olfr1506
Genomic Map