siRNA Details
| Gene Symbol: Olfr1269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146342.1 | NP_666454.1 | 1 | 660 | ||
| GenBank mRNA | BC116956.1 | - | 1 | 681 | 40126275 | |
| BC116958.1 | - | 1 | 681 | 40126277 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1269| Gene Symbol: Olfr1269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146342.1 | NP_666454.1 | 1 | 660 | ||
| GenBank mRNA | BC116956.1 | - | 1 | 681 | 40126275 | |
| BC116958.1 | - | 1 | 681 | 40126277 | ||