siRNA Details
Gene Symbol: Olfr1133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146351.2 | NP_666463.2 | 1 | 202 | ||
GenBank mRNA | BC104086.1 | - | 1 | 202 | ||
BC104087.2 | - | 1 | 201 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146351.2 | NP_666463.2 | 1 | 202 | ||
GenBank mRNA | BC104086.1 | - | 1 | 202 | ||
BC104087.2 | - | 1 | 201 |