siRNA Details
| Gene Symbol: Olfr976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146367.2 | NP_666479.1 | 1 | 208 | ||
| GenBank mRNA | AK135940.1 | - | 2 | 305 | ||
| BC119350.1 | - | 1 | 208 | 8734099 | ||
| BC120621.1 | - | 1 | 208 | 40129544 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr976| Gene Symbol: Olfr976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146367.2 | NP_666479.1 | 1 | 208 | ||
| GenBank mRNA | AK135940.1 | - | 2 | 305 | ||
| BC119350.1 | - | 1 | 208 | 8734099 | ||
| BC120621.1 | - | 1 | 208 | 40129544 | ||