siRNA Details
| Gene Symbol: Olfr361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146368.1 | NP_666480.1 | 1 | 524 | ||
| GenBank mRNA | BC104114.1 | - | 1 | 522 | ||
| BC104115.1 | - | 1 | 522 | |||
| BC114393.1 | - | 1 | 523 | |||
| BC114424.1 | - | 1 | 523 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr361| Gene Symbol: Olfr361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146368.1 | NP_666480.1 | 1 | 524 | ||
| GenBank mRNA | BC104114.1 | - | 1 | 522 | ||
| BC104115.1 | - | 1 | 522 | |||
| BC114393.1 | - | 1 | 523 | |||
| BC114424.1 | - | 1 | 523 | |||