siRNA Details
| Gene Symbol: Olfr434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146369.1 | NP_666481.1 | 1 | 809 | ||
| GenBank mRNA | BC120611.1 | - | 1 | 841 | 40129534 | |
| BC120613.1 | - | 1 | 841 | 40129536 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr434| Gene Symbol: Olfr434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146369.1 | NP_666481.1 | 1 | 809 | ||
| GenBank mRNA | BC120611.1 | - | 1 | 841 | 40129534 | |
| BC120613.1 | - | 1 | 841 | 40129536 | ||