siRNA Details
| Gene Symbol: Olfr654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146379.1 | NP_666491.1 | 1 | 108 | ||
| GenBank mRNA | BC125556.1 | - | 1 | 144 | 40130071 | |
| BC125558.1 | - | 1 | 144 | 40130073 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr654| Gene Symbol: Olfr654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146379.1 | NP_666491.1 | 1 | 108 | ||
| GenBank mRNA | BC125556.1 | - | 1 | 144 | 40130071 | |
| BC125558.1 | - | 1 | 144 | 40130073 | ||