siRNA Details
| Gene Symbol: Olfr460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146383.1 | NP_666495.1 | 1 | 400 | ||
| GenBank mRNA | BC119048.1 | - | 1 | 431 | 8733797 | |
| BC120738.1 | - | 1 | 431 | 40129661 | ||
More Information
Other Species Targeted
Set Membership: |
|

Olfr460
Genomic Map