siRNA Details
| Gene Symbol: Olfr1347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146385.1 | NP_666497.1 | 1 | 74 | ||
| GenBank mRNA | BC128003.1 | - | 1 | 78 | 40135936 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1347| Gene Symbol: Olfr1347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146385.1 | NP_666497.1 | 1 | 74 | ||
| GenBank mRNA | BC128003.1 | - | 1 | 78 | 40135936 | |
Set Membership: |
|