siRNA Details
| Gene Symbol: Olfr1006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146570.2 | NP_666781.2 | 2 | 177 | ||
| GenBank mRNA | DR065595.1 | - | 204 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1006| Gene Symbol: Olfr1006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146570.2 | NP_666781.2 | 2 | 177 | ||
| GenBank mRNA | DR065595.1 | - | 204 | |||