siRNA Details
| Gene Symbol: Olfr1032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146579.2 | NP_666790.2 | 1 | 912 | ||
| GenBank mRNA | BC113200.1 | - | 1 | 911 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1032| Gene Symbol: Olfr1032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146579.2 | NP_666790.2 | 1 | 912 | ||
| GenBank mRNA | BC113200.1 | - | 1 | 911 | ||