siRNA Details
| Gene Symbol: Olfr1030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146588.2 | NP_666799.1 | 2 | 180 | ||
| GenBank mRNA | BC128001.1 | - | 1 | 151 | 40135864 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1030| Gene Symbol: Olfr1030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146588.2 | NP_666799.1 | 2 | 180 | ||
| GenBank mRNA | BC128001.1 | - | 1 | 151 | 40135864 | |