siRNA Details
| Gene Symbol: Olfr521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146356.2 | NP_666468.2 | 1 | 947 | ||
| GenBank mRNA | BC106825.1 | - | 1 | 945 | ||
| BC106826.1 | - | 1 | 947 | 40044016 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr521| Gene Symbol: Olfr521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146356.2 | NP_666468.2 | 1 | 947 | ||
| GenBank mRNA | BC106825.1 | - | 1 | 945 | ||
| BC106826.1 | - | 1 | 947 | 40044016 | ||