siRNA Details
| Gene Symbol: Olfr1093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146366.1 | NP_666478.1 | 1 | 748 | ||
| GenBank mRNA | BC138167.1 | - | 1 | 835 | 8861186 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1093| Gene Symbol: Olfr1093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146366.1 | NP_666478.1 | 1 | 748 | ||
| GenBank mRNA | BC138167.1 | - | 1 | 835 | 8861186 | |
Set Membership: |
|