siRNA Details
| Gene Symbol: Olfr509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146372.1 | NP_666484.1 | 1 | 698 | ||
| GenBank mRNA | BC132049.1 | - | 1 | 782 | 40130326 | |
| BC132435.1 | - | 1 | 782 | 40130712 | ||
More Information
Set Membership: |
|

Olfr509
Genomic Map