siRNA Details
Gene Symbol: Olfr460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146383.1 | NP_666495.1 | 1 | 552 | ||
GenBank mRNA | BC119048.1 | - | 1 | 583 | 8733797 | |
BC120738.1 | - | 1 | 583 | 40129661 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
