siRNA Details
| Gene Symbol: Olfr862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146562.1 | NP_666773.1 | 1 | 604 | ||
| GenBank mRNA | BC120769.1 | - | 1 | 656 | 40129692 | |
| BC125340.1 | - | 1 | 656 | 40129855 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr862| Gene Symbol: Olfr862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146562.1 | NP_666773.1 | 1 | 604 | ||
| GenBank mRNA | BC120769.1 | - | 1 | 656 | 40129692 | |
| BC125340.1 | - | 1 | 656 | 40129855 | ||