siRNA Details
| Gene Symbol: Olfr1046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146582.2 | NP_666793.2 | 1 | 914 | ||
| GenBank mRNA | BC099891.1 | - | 1 | 913 | ||
| BC099892.1 | - | 1 | 913 | |||
| BC103540.1 | - | 1 | 913 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1046| Gene Symbol: Olfr1046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146582.2 | NP_666793.2 | 1 | 914 | ||
| GenBank mRNA | BC099891.1 | - | 1 | 913 | ||
| BC099892.1 | - | 1 | 913 | |||
| BC103540.1 | - | 1 | 913 | |||