siRNA Details
| Gene Symbol: Olfr1026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146584.2 | NP_666795.2 | 1 | 819 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1026| Gene Symbol: Olfr1026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146584.2 | NP_666795.2 | 1 | 819 | ||