siRNA Details
Gene Symbol: Olfr168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146357.1 | NP_666469.1 | 1 | 915 | ||
GenBank mRNA | BC127969.1 | - | 1 | 919 | 40135070 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146357.1 | NP_666469.1 | 1 | 915 | ||
GenBank mRNA | BC127969.1 | - | 1 | 919 | 40135070 |