siRNA Details
| Gene Symbol: Olfr557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146361.2 | NP_666473.2 | 1 | 967 | ||
| GenBank mRNA | BC145899.1 | - | 1 | 967 | 40131202 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr557| Gene Symbol: Olfr557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146361.2 | NP_666473.2 | 1 | 967 | ||
| GenBank mRNA | BC145899.1 | - | 1 | 967 | 40131202 | |