siRNA Details
| Gene Symbol: Olfr1012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146568.2 | NP_666779.1 | 1 | 459 | ||
| GenBank mRNA | BC111581.1 | - | 1 | 457 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1012| Gene Symbol: Olfr1012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146568.2 | NP_666779.1 | 1 | 459 | ||
| GenBank mRNA | BC111581.1 | - | 1 | 457 | ||