siRNA Details
| Gene Symbol: Olfr1009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146572.2 | NP_666783.1 | 1 | 886 | ||
| GenBank mRNA | BC111578.1 | - | 1 | 885 | ||
| BC148238.1 | - | 1 | 886 | 40042413 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1009| Gene Symbol: Olfr1009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146572.2 | NP_666783.1 | 1 | 886 | ||
| GenBank mRNA | BC111578.1 | - | 1 | 885 | ||
| BC148238.1 | - | 1 | 886 | 40042413 | ||