siRNA Details
Gene Symbol: Olfr502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146739.1 | NP_666950.1 | 1 | 434 | ||
GenBank mRNA | BC119235.1 | - | 1 | 453 | 8733984 | |
BC120678.1 | - | 1 | 453 | 40129601 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146739.1 | NP_666950.1 | 1 | 434 | ||
GenBank mRNA | BC119235.1 | - | 1 | 453 | 8733984 | |
BC120678.1 | - | 1 | 453 | 40129601 |